missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GDNF Monoclonal Antibody (K92005_4F10), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA555725
Description
Sequence of this protein is as follows: RNRQAAAANP ENSRGKGRRG QRGKNRGCVL TAIHLNVTDL GLGYETKEEL IFRYCSGSCD AAETTYDKIL KNLSRNRRLV
GDNF is a glycosylated, disulfide-bonded homodimer molecule. It was first discovered as a potent survival factor for midbrain dopaminergic neurons and was then shown to rescue these neurons in animal models of Parkinson's disease. GDNF is about 100 times more efficient survival factor for spinal motor neurons than the neurotrophins. GDNF is a neurotrophic factor that enhances survival and morphological differentiation of dopaminergic neurons and increases their high-affinity dopamine uptake.Chemical Identifiers
| GDNF | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| AI385739; Astrocyte-derived trophic factor; ATF; ATF1; ATF2; Gdnf; glial cell derived neurotrophic factor; glial cell line derived neurotrophic factor; Glial cell line derived neutrophic factor; glial cell line-derived neurotrophic factor; gndf; HFB1-GDNF; hGDNF; H-GDNF; HSCR3; mGDNF; neurotrophic factor | |
| Recombinant human GDNF (immunogen range is 91-170). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| GDNF |
| K92005_4F10 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P39905 | |
| 2668 | |
| IgG1 | |
| Protein A | |
| Antibody |
Specifications
| GDNF | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P39905 | |
| GDNF | |
| Recombinant human GDNF (immunogen range is 91-170). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| K92005_4F10 | |
| Unconjugated | |
| GDNF | |
| AI385739; Astrocyte-derived trophic factor; ATF; ATF1; ATF2; Gdnf; glial cell derived neurotrophic factor; glial cell line derived neurotrophic factor; Glial cell line derived neutrophic factor; glial cell line-derived neurotrophic factor; gndf; HFB1-GDNF; hGDNF; H-GDNF; HSCR3; mGDNF; neurotrophic factor | |
| Mouse | |
| Protein A | |
| RUO | |
| 2668 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |