missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GC Syringes for Agilent Instruments

Catalog No. 03251154
Change view
Click to view available options
Volume (Metric):
5 μL
10 μL
Description:
Straight
Tapered
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Description
03251154 10 μL Tapered
03251150 5 μL Straight
03251151 10 μL Straight
03251152 10 μL Straight
03251153 10 μL Tapered
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 03251154 Supplier Thermo Scientific™ Supplier No. 365D1636

Easily accomplish accurate, precise, and reproducible injections with Thermo Scientific™ GC Syringes for Agilent Instruments.

Achieve accurate, precise and reproducible injections easily using Thermo Scientific™ GC Syringes designed specifically for Agilent™ GC systems. Optimized for septum piercing, the rough, field-tested syringes are made of borosilicate glass with metal flanges of either stainless steel or nickel-plated brass.

  • Accurate, precise and reproducible over a range of volumes
  • Fixed needle syringes have a maximum temperature range of 5° to 70°C
  • 5 to 100μL capacity needles have a maximum safe working pressure of 1000psi; 100psi maximum functional pressure
  • Cone point needle tip to prevent plunger binding with dirty samples
  • 42mm length

Specifications

Description Tapered
Needle Gauge 23 to 26s G
Length (Metric) 42 mm
Product Type GC Autosampler Syringe, Fixed Needle
Disposable Reusable
Quantity 6 Pk.
Volume (Metric) 10 μL
Length (Metric) Needle 42 mm
Needle Point Style Cone
For Use With (Equipment) Agilent GC Instruments

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.