missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GC SMART Syringes for TriPlus™ RSH SMART Autosampler

Catalog No. 03050915
Change view
Click to view available options
Volume (Metric):
0.5 μL
1 μL
5 μL
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
1 mL
2.5 mL
5 mL
10 mL
Needle Gauge:
23s G
26s G
19 G
22 G
23 G
26 G
Length (Metric) Needle:
57 mm
65 mm
85 mm
Product Type:
Gas-tight GC Autosampler Syringe, Cone
Gas-tight GC Autosampler Syringe, LC
Gas-tight GC Autosampler Syringe, Side Hole
GC Autosampler Syringe, Bevel
GC Autosampler Syringe, Cone
38 product options available for selection
Product selection table with 38 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Needle Gauge Length (Metric) Needle Product Type
03050915 5 μL 26s G 57 mm GC Autosampler Syringe, Cone
03050910 0.5 μL 23 G 57 mm GC Autosampler Syringe, Cone
03050911 1 μL 23 G 57 mm GC Autosampler Syringe, Cone
03050912 5 μL 23s G 85 mm GC Autosampler Syringe, Cone
03050913 5 μL 23s G 57 mm GC Autosampler Syringe, Cone
03050914 5 μL 26s G 85 mm GC Autosampler Syringe, Cone
03050916 10 μL 23s G 85 mm GC Autosampler Syringe, Cone
03050917 10 μL 23s G 57 mm GC Autosampler Syringe, Cone
03050918 10 μL 26s G 85 mm GC Autosampler Syringe, Cone
03050919 10 μL 26s G 57 mm GC Autosampler Syringe, Cone
03050920 10 μL 23s G 85 mm Gas-tight GC Autosampler Syringe, Cone
03050921 10 μL 23s G 57 mm Gas-tight GC Autosampler Syringe, Cone
03050922 10 μL 26s G 85 mm Gas-tight GC Autosampler Syringe, Cone
03254198 10 μL 26s G 57 mm Gas-tight GC Autosampler Syringe, Cone
03050923 10 μL 26s G 57 mm GC Autosampler Syringe, Bevel
03050924 25 μL 23s G 57 mm Gas-tight GC Autosampler Syringe, Cone
03050925 25 μL 26s G 85 mm Gas-tight GC Autosampler Syringe, Cone
03050926 25 μL 26s G 57 mm Gas-tight GC Autosampler Syringe, Cone
03050927 50 μL 23s G 57 mm Gas-tight GC Autosampler Syringe, Cone
03050928 50 μL 26s G 85 mm Gas-tight GC Autosampler Syringe, Cone
03050929 50 μL 26s G 57 mm Gas-tight GC Autosampler Syringe, Cone
03050930 100 μL 23s G 57 mm Gas-tight GC Autosampler Syringe, Cone
03050931 100 μL 26s G 85 mm Gas-tight GC Autosampler Syringe, Cone
03050932 100 μL 26s G 57 mm Gas-tight GC Autosampler Syringe, Cone
03050933 100 μL 23 G 85 mm Gas-tight GC Autosampler Syringe, Side Hole
03050934 100 μL 23 G 57 mm Gas-tight GC Autosampler Syringe, Side Hole
03050935 250 μL 26 G 85 mm Gas-tight GC Autosampler Syringe, Cone
03050936 250 μL 26 G 57 mm Gas-tight GC Autosampler Syringe, Cone
03050937 250 μL 23 G 85 mm Gas-tight GC Autosampler Syringe, Side Hole
03050938 250 μL 23 G 57 mm Gas-tight GC Autosampler Syringe, Side Hole
03050939 500 μL 26 G 85 mm Gas-tight GC Autosampler Syringe, Cone
03050940 500 μL 26 G 57 mm Gas-tight GC Autosampler Syringe, Cone
03050941 500 μL 23 G 57 mm Gas-tight GC Autosampler Syringe, Side Hole
03050942 1 mL 22 G 57 mm Gas-tight GC Autosampler Syringe, LC
03050943 1 mL 23 G 65 mm Gas-tight GC Autosampler Syringe, Side Hole
03050944 2.5 mL 23 G 65 mm Gas-tight GC Autosampler Syringe, Side Hole
03050945 5 mL 23 G 65 mm Gas-tight GC Autosampler Syringe, Side Hole
03050946 10 mL 19 G 57 mm Gas-tight GC Autosampler Syringe, LC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 38 options available.
38 options
Catalog No. 03050915 Supplier Thermo Scientific™ Supplier No. 365C0251SM
Only null left

Use these GC syringes embedded with an ID chip to track part and lot numbers, usage, and history to increase data confidence and ensure system compliance.

Use the unique technology of Thermo Scientific™ GC SMART Syringes as a traceable usage-based approach to gas chromatography (GC) syringe management, resulting in increased reliability, instrument up-time, confidence in the results, and full traceability. When installed on a Thermo Scientific™ TriPlus™ RSH SMART autosampler, the SMART syringes benefit from the unique SMART technology. Each syringe is equipped with an ID chip, which stores information such as part number, lot number, usage range, and history for that specific syringe. The information stored on the chip can be directly accessed and reported via Thermo Scientific™ Chromeleon™ CDS software.

SMART syringes are compatible with all TriPlus™ RSH instruments as well as PAL platforms and AOC-6000 series instruments.

SMART Technology Benefits
- Gain higher confidence in analytical results
- Ensure error free usage of syringes through automatic identification
- Support GLP accreditation with usage history traceability
- Optimize consumption by monitoring of usage conditions and lifetime
- Facilitate syringe management thanks to color-coded health notification dashboard integrated in Chromeleon™ CDS

Specifications

Description 5 μL FN, 26 s G, Cone, 57 mm
Needle Gauge 26s G
Product Type GC Autosampler Syringe, Cone
Volume (Metric) 5 μL
Length (Metric) Needle 57 mm
Needle Point Style Cone
For Use With (Equipment) TriPlus RSH SMART Autosampler, All TriPlus RSH Instruments and All PAL3 Models