missing translation for 'onlineSavingsMsg'
Learn More

GBR 12909 dihydrochloride, Thermo Scientific Chemicals

Catalog No. AAJ60684MA
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60684MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ60684MA Supplier Thermo Scientific Chemicals Supplier No. J60684MA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Dopamine uptake inhibitor

Dopamine uptake inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 67469-78-7
Molecular Formula C28H34Cl2F2N2O
Molecular Weight (g/mol) 523.49
MDL Number MFCD00055193
InChI Key MIBSKSYCRFWIRU-UHFFFAOYSA-N
PubChem CID 104920
ChEBI CHEBI:64086
IUPAC Name 1-[2-[bis(4-fluorophenyl)methoxy]ethyl]-4-(3-phenylpropyl)piperazine;dihydrochloride
SMILES C1CN(CCN1CCCC2=CC=CC=C2)CCOC(C3=CC=C(C=C3)F)C4=CC=C(C=C4)F.Cl.Cl

Specifications

Color White
Quantity 10 mg
Synonym vanoxerine dihydrochloride, gbr 12909 dihydrochloride, vanoxeamine, gbr-12909 dihydrochloride, vanoxerine hydrochloride, unii-mwo1ip03ev, mwo1ip03ev, vanoxerine dihydrochloride gbr12909, gbr12909 dihydrochloride, 1-2-bis 4-fluorophenyl methoxy ethyl-4-3-phenylpropyl piperazine dihydrochloride
Solubility Information Soluble in DMSO to 50mM; Soluble to 5mM in water with warming.
Formula Weight 523.49
Physical Form Powder
Chemical Name or Material GBR 12909 dihydrochloride
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only