missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Gauge Marking Labels

Color-coded labels to mark gauges to easily identify acceptable or dangerous levels. Brady Gauge Marking Label are transparent and available in size 9 x 10 in.

Supplier:  Brady™ 30697

 View more versions of this product

Catalog No. 19-110-830


Only null left
Explore available promotions

Description

Description

  • To easily identify acceptable or dangerous levels
  • Made of reflective polyester sheeting
  • Color-coded and available in size 228.6W x 254mm H
Specifications

Specifications

Abrasion, chemical and weather resistant
Red
Reflective Polyester Sheeting
Rectangle
228.6 x 254 mm (W x H)
Acrylic, Permanent
Gloss
Gauge marking label
9 x 10 in. (W x H)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions