Learn More
Cytiva GammaBind Plus Sepharose
Binds to all IgG subclasses. Cytiva GammaBind Plus Sepharose is used for lab-scale affinity purification of IgG from a large variety of species and has better binding to rat and mouse IgG subclasses compared to standard GammaBind Sepharose.
Supplier: Cytiva 17088601
Description
GammaBind Plus Sepharose may be used to analyze and purify classes, subclasses and fragments of immunoglobulins from any biological fluid or cell culture medium.- Recombinant Protein G type 3 coupled to Sepharose CL-6B for enhanced binding to rat and mouse IgG subclasses at laboratory scale
- Binds to all IgG subclasses from human, mouse, and rat; binds total IgG from guinea pig, rabbit, goat, cow, sheep, and horse
- Yields up to 18mg/ml mouse IgG
- Available in 5ml and 25ml sizes
Specifications
| >130 cm/h | |
| 90 μm, d50v is the median particle size of the cumulative volume distribution. | |
| 5 mL |
| Recombinant GammaBind G type 3 lacking albumin-binding region | |
| 2°C to 8°C, 20% Ethanol |