missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva GammaBind Plus Sepharose
Description
GammaBind Plus Sepharose may be used to analyze and purify classes, subclasses and fragments of immunoglobulins from any biological fluid or cell culture medium.
- Recombinant Protein G type 3 coupled to Sepharose CL-6B for enhanced binding to rat and mouse IgG subclasses at laboratory scale
- Binds to all IgG subclasses from human, mouse, and rat; binds total IgG from guinea pig, rabbit, goat, cow, sheep, and horse
- Yields up to 18mg/ml mouse IgG
- Available in 5ml and 25ml sizes
Specifications
Specifications
| Flow Rate | >130 cm/h |
| Ligand | Recombinant GammaBind G type 3 lacking albumin-binding region |
| Particle Size | 90 μm, d50v is the median particle size of the cumulative volume distribution. |
| Content And Storage | 2°C to 8°C, 20% Ethanol |
| Quantity | 25 mL |