missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma-Methyl-gamma-decanolactone 98.0+%, TCI America™
$115.57 - $728.84
Chemical Identifiers
| CAS | 7011-83-8 |
|---|---|
| Molecular Formula | C11H20O2 |
| Molecular Weight (g/mol) | 184.279 |
| MDL Number | MFCD00036642 |
| InChI Key | ALWUKGXLBSQSMA-UHFFFAOYSA-N |
| Synonym | gamma-Methyl-gamma-decalactone |
| PubChem CID | 285097 |
| IUPAC Name | 5-hexyl-5-methyloxolan-2-one |
| SMILES | CCCCCCC1(CCC(=O)O1)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M078625ML
|
TCI America
M078625ML |
25 mL |
Each for $115.57
|
|
|||||
|
M0786500ML
|
TCI America
M0786500ML |
500 mL |
Each for $728.84
|
|
|||||
Chemical Identifiers
| 7011-83-8 | |
| 184.279 | |
| ALWUKGXLBSQSMA-UHFFFAOYSA-N | |
| 285097 | |
| CCCCCCC1(CCC(=O)O1)C |
| C11H20O2 | |
| MFCD00036642 | |
| gamma-Methyl-gamma-decalactone | |
| 5-hexyl-5-methyloxolan-2-one |
Specifications
| 7011-83-8 | |
| 160°C | |
| MFCD00036642 | |
| gamma-Methyl-gamma-decalactone | |
| CCCCCCC1(CCC(=O)O1)C | |
| 184.279 | |
| 184.28 | |
| Liquid |
| Yellow | |
| C11H20O2 | |
| 25 mL | |
| ALWUKGXLBSQSMA-UHFFFAOYSA-N | |
| 5-hexyl-5-methyloxolan-2-one | |
| 285097 | |
| ≥98.0% (GC) | |
| gamma-Methyl-gamma-decanolactone |
Safety and Handling
EINECSNumber : (9)-0137
RTECSNumber : HD9800000
TSCA : Yes