missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma-Methyl-gamma-decanolactone 98.0+%, TCI America™
Supplier: TCI America M0786500ML
Specifications
| gamma-Methyl-gamma-decanolactone | |
| Yellow | |
| C11H20O2 | |
| 500 mL | |
| ALWUKGXLBSQSMA-UHFFFAOYSA-N | |
| 5-hexyl-5-methyloxolan-2-one | |
| 285097 | |
| ≥98.0% (GC) |
| 7011-83-8 | |
| 160°C | |
| MFCD00036642 | |
| gamma-Methyl-gamma-decalactone | |
| CCCCCCC1(CCC(=O)O1)C | |
| 184.279 | |
| 184.28 | |
| Liquid |
Chemical Identifiers
| 7011-83-8 | |
| 184.279 | |
| ALWUKGXLBSQSMA-UHFFFAOYSA-N | |
| 285097 | |
| CCCCCCC1(CCC(=O)O1)C |
| C11H20O2 | |
| MFCD00036642 | |
| gamma-Methyl-gamma-decalactone | |
| 5-hexyl-5-methyloxolan-2-one |
Safety and Handling
EINECSNumber : (9)-0137
RTECSNumber : HD9800000
TSCA : Yes