missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Galacto-Star™ β-Galactosidase Reporter Gene Assay System for Mammalian Cells

Catalog No. T1012
Encompass
Change view
Click to view available options
Quantity:
1800 Assays
600 Assays
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1012 600 Assays
T1014 1800 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T1012 Supplier Invitrogen™ Supplier No. T1012

Includes

Lysis solution, substrate, enhancer, and required buffer

The Galacto-Star™ One-Step β-Galactosidase Reporter Gene Assay System chemiluminescent reporter gene assay system enables rapid, sensitive detection of β-galactosidase in mammalian cell lysates. SensitiveThe chemiluminescent assay exhibits over three o

  • Glow assay kinetics provides a window during which measurements may be performed, facilitating HTS applications where assay automation is used
  • Can be used with either mammalian or yeast model systems, providing flexibility in choice of model systems
  • Wide dynamic range of β-galactosidase assay lets the user measure enzyme level accurately from femtogram to nanogram range
  • Assay sensitivity is 100- to 1,000-fold better than either the isotopic/non-isotopic assays for chloramphenicol acetyl transferase (CAT) or the colorimetric/fluorescent assays for β-galactosidase, providing greater sensitivity than competing assays
  • Highly sensitive assay with a wide dynamic range permits detection of high and low levels of reporter without performing numerous sample dilutions
  • Non-radioactive reporter gene assay kit eliminates concerns over use of radioisotopes
  • Assay can be completed in about one hour, providing fast assay turnaround

Sensitive
The chemiluminescent assay exhibits over three orders of magnitude greater sensitivity than colorimetric β-galactosidase assays and can be completed in a fraction of the time required for assays for chloramphenicol acetyltransferase (CAT) and ELISA-based reporter assays.

Versatile
The Galacto-Star system includes Galacton-Star chemiluminescent substrate for β-galactosidase and is suited for measurement in both luminometers and scintillation counters. Automatic injection is not required.

Long-Lasting Signal
Light emission from the Galacto-Star assay reaches maximum in 60 to 90 minutes and remains constant for nearly one hour. Cell lysate is mixed with reaction buffer containing Galacton-Star substrate and Sapphire-II™ luminescence enhancer. After a 60-90 minute incubation at room temperature, the signal is measured in a luminometer or scintillation counter.

Specifications

  • Assay Type: β-Gal (lacZ) Assay
  • Compatible Cells: Mammalian Cells
  • Detection Method: Chemiluminescent
  • For Use With Equipment: Luminometer (Microplate)
  • Form: Liquid
  • Format: 96-well plate
  • High Throughput Compatibility: High Throughput-Compatible
  • Readout: End Point
  • Sample Type (Specific): Cell Cultures
  • Substrate: Galacton-Star
  • Substrate Properties: Chemical Substrate
  • Substrate Type: Beta-Gal Substrate
  • Target Enzyme: Beta-Galactosidase

Specifications

Product Type β-Galactosidase Reporter Gene Assay System
Detection Method Chemiluminescence
For Use With (Equipment) Luminometer (Microplate)
Assay β-Gal (lacZ) Assay
Content And Storage This kit contains the lysis solution, substrate, enhancer, and required buffer. Store the product at 4°C.
Compatible Cells Mammalian Cells
High-throughput Compatibility High-throughput Compatible
Label or Dye Unconjugated
Readout End Point
Sample Type Cell Cultures
Substrate Galacton-Star™
Substrate Type Beta-Gal Substrate
Substrate Properties Chemical Substrate
Target Enzyme Beta-Galactosidase
Product Line Galacto-Star, NovaBright
For Use With (Application) Enzyme Assay
Quantity 600 Assays
Form Liquid
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.