missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Galacto-Light™ Reaction Buffer Diluent with Galacton-Plus™ Substrate

Catalog No. T1055
Encompass
Change view
Click to view available options
Quantity:
1800 assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1055 1800 assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1055 Supplier Invitrogen™ Supplier No. T1055

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Concentrated substrate and dilution buffer

Established system with extended light emission and high sensitivity for reporter gene assays. Thermo Scientific Galacto-Light Reaction Buffer Diluent and the Galacton-Plus chemiluminescent substrate solution are used with the Galacto-Light Plus Reporter

The Galacto-Light™ Reaction Buffer Diluent and the Galacton-Plus™ chemiluminescent substrate solution are used with the Galacto-Light™ Plus Reporter Gene Assay System.

Use With

Light Emission Accelerator-II

For Research Use Only. Not for use in diagnostics procedures.

Specifications

Product Type ELISA Substrate and Diluent
Quantity 1800 assays
Detection Method Chemiluminescence
Conjugate Unconjugated
Form Liquid
Format 96-well Plates
For Use With (Application) Enzyme Assay
For Use With (Equipment) Luminometer (Microplate)
High-throughput Compatibility High-throughput Compatible
Product Line Galacto-Light, Galacton-Plus
Sample Type Cell Cultures
Substrate Properties Chemical Substrate
Substrate Type Beta-Gal
Content And Storage This kit contains the concentrated substrate and the dilution buffer.
Store the product at 4°C.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.