missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Galacto-Light Plus™ β-Galactosidase Reporter Gene Assay System DFS Item

Rapid, ultrasensitive detection of β-galactosidase in reporter gene assays. Thermo Scientific Galacto-Light Plus β-Galactosidase Reporter Gene Assay System is designed for luminometers without injectors.

$502.00 - $7630.00

Specifications

Product Type β-Galactosidase Reporter Gene Assay System
Detection Method Chemiluminescence
For Use With (Equipment) Luminometer (Microplate)
Assay β-Gal (lacZ) Assay
Content And Storage This kit contains the lysis solution, substrate, reaction buffer, and accelerator.
Store at 4°C.
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T1007 DFS Item
View Documents
Invitrogen™
T1007
200 Assays
Each for $502.00
Only null left
 
T1009 DFS Item
View Documents
Invitrogen™
T1009
15,000 Assays
Each for $7,630.00
Only null left
 
T1011 DFS Item
View Documents
Invitrogen™
LST1011
600 Assays
Each for $996.00
Only null left
 
Description

Description

  • Wide dynamic range of β-galactosidase assay lets the user measure enzyme level accurately from femtogram to nanogram range
  • Assay sensitivity is 100- to 1000-fold better than with either isotopic/non-isotopic assays for chloramphenicol acetyl transferase (CAT) or colorimetric/fluorescent assays for β-galactosidase, providing greater sensitivity than competing assays
  • Highly sensitive assay with wide dynamic range permits detection of high and low levels of reporter without performing numerous sample dilutions
  • Non-radioactive reporter gene assay kit eliminates concerns over use of radioisotopes
  • Assay can be completed in about one hour, providing fast assay turnaround

Sensitive
This chemiluminescent assay exhibits over three orders of magnitude greater sensitivity compared to colorimetric β-galactosidase assays and can be completed in a fraction of time required for assays for chloramphenicol acetyltransferase (CAT) and ELISA-based reporter assays.

Ready-to-Use
The Galacto-Light Plus system incorporates chemiluminescent substrates, reaction buffer, and light-emission accelerator containing luminescence enhancers, in a ready-to-use format. The reaction buffer is supplied at a pH that aids in discrimination of endogenous β-galactosidase, while having little effect on the bacterial form of the enzyme. This enables sensitive detection even in cell lines with relatively high levels of endogenous β-galactosidase activity.

Specifications

  • Assay Type: β-Gal (lacZ) Assay
  • Compatible Cells: Mammalian Cells
  • Detection Method: Chemiluminescent
  • For Use With Equipment: Luminometer (Microplate)
  • Form: Liquid
  • Format: 96-well plate
  • High Throughput Compatibility: High Throughput-Compatible
  • Readout: End Point
  • Sample Type (Specific): Cell Cultures
  • Substrate: Galacton-Plus
  • Substrate Properties: Chemical Substrate
  • Substrate Type: Beta-Gal Substrate
  • Target Enzyme: Beta-Galactosidase
Specifications

Specifications

β-Galactosidase Reporter Gene Assay System
Luminometer (Microplate)
This kit contains the lysis solution, substrate, reaction buffer, and accelerator.
Store at 4°C.
High-throughput Compatible
End Point
Galacton-Plus™
Chemical Substrate
Enzyme Assay
Chemiluminescence
β-Gal (lacZ) Assay
Mammalian Cells
Unconjugated
Cell Cultures
Beta-Gal Substrate
Beta-Galactosidase
Liquid
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.