missing translation for 'onlineSavingsMsg'
Learn More

Galactitol 98.0+%, TCI America™

Supplier:  TCI America G0005250G

Quantity 250 g

 View more versions of this product

Catalog No. G0005250G


Only null left

Description

Description

Specifications

Chemical Identifiers

608-66-2
182.172
FBPFZTCFMRRESA-GUCUJZIJSA-N
11850
(2R,3S,4R,5S)-hexane-1,2,3,4,5,6-hexol
C6H14O6
MFCD00064288
dulcitol, galactitol, dulcite, d-galactitol, dulcose, euonymit, melampyrin, melampyrit, d-dulcitol, 2r,3s,4r,5s-hexane-1,2,3,4,5,6-hexaol
CHEBI:16813
C(C(C(C(C(CO)O)O)O)O)O

Specifications

Galactitol
189°C
280°C
250 g
dulcitol, galactitol, dulcite, d-galactitol, dulcose, euonymit, melampyrin, melampyrit, d-dulcitol, 2r,3s,4r,5s-hexane-1,2,3,4,5,6-hexaol
C(C(C(C(C(CO)O)O)O)O)O
182.172
CHEBI:16813
≥98.0% (GC)
608-66-2
White
C6H14O6
MFCD00064288
FBPFZTCFMRRESA-GUCUJZIJSA-N
(2R,3S,4R,5S)-hexane-1,2,3,4,5,6-hexol
11850
182.17
Crystalline Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions