accessibility menu, dialog, popup

UserName

HOOHOHOHOHOH

CAS RN 608-66-2

IUPAC Name: hexane-1,2,3,4,5,6-hexol
Synonyms: mannitol sorbo sorbitol D(-)-mannitol 甘露醇
Molecular Weight (g/mol): 182.17
Molecular Formula: C6H14O6
InChi Key: FBPFZTCFMRRESA-UHFFFAOYNA-N
SMILES: OCC(O)C(O)C(O)C(O)CO
Boiling Point 
  • (3)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)