missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Gal-Screen™ β-Galactosidase Reporter Gene Assay System for Mammalian Cells

Catalog No. T1029
Encompass
Change view
Click to view available options
Quantity:
10,000 Assays
1000 Assays
200 Assays
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1029 200 Assays
T1027 1000 Assays
T1028 10,000 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T1029 Supplier Invitrogen™ Supplier No. T1029

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Enzyme substrate and reaction buffer

Combines direct cell lysis with rapid, ultra-sensitive chemiluminescent detection of β-galactosidase reporter enzyme.

The Gal-Screen™ assay system combines direct cell lysis with rapid, ultra-sensitive chemiluminescent detection of β-galactosidase reporter enzyme.

  • Homogeneous assay format allows detection of β-galactosidase in the presence of normal culture media without removal of media and without an additional cell lysis step, providing the easiest and most streamlined assay procedure possible.
  • The kinetics of the glow-assay provides a 'window' during which measurements may be performed, facilitating HTS applications where assay automation is used.
  • Can be used with either mammalian or yeast model systems, providing flexibility in choice of model systems.
  • Wide dynamic range of β-galactosidase assay enables accurate measurement of enzyme from femtogram to nanogram range.
  • Assay sensitivity is 100 to 1,000-fold better than either the isotopic/non-isotopic assays for chloramphenicol acetyl transferase (CAT) or the colorimetric/fluorescent assays for β-galactosidase, providing greater sensitivity than competing assay technologies.
  • Highly sensitive assay with a wide dynamic range permits detection of high and low levels of reporter without performing numerous sample dilutions.
  • Non-radioactive reporter gene assay kit eliminates concerns over use of radioisotopes.
  • Assay can be completed in about one hour, providing fast assay turnaround.

Ideal for Screening

This homogeneous assay is ideally suited for screening applications where assay automation is required. The Gal-Screen™ system uses Galacton-Star™ chemiluminescent substrate for convenient measurement in a luminometer. Light emission reaches maximum in 60-90 minutes and remains constant for 45-90 minutes.

For Research Use Only. Not for use in diagnostics procedures.

Specifications

Product Type β-Galactosidase Reporter Gene Assay System
Detection Method Chemiluminescence
For Use With (Equipment) Luminometer
Assay β-Gal (lacZ) Assay
Content And Storage This kit contains the enzyme substrate and the reaction buffer.
Store the product at 4°C.
Compatible Cells Mammalian Cells
High-throughput Compatibility High-throughput Compatible
Label or Dye Unconjugated
Readout End Point
Sample Type Cell Cultures
Substrate Galacton-Star™
Substrate Type Beta-Gal Substrate
Substrate Properties Chemical Substrate
Target Enzyme Beta-Galactosidase
Product Line Gal-Screen, NovaBright
For Use With (Application) Enzyme Assay
Quantity 200 Assays
Form Liquid
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.