Learn More
Gadolinium(III) oxide, 99.9%, (trace metal basis)
CAS: 12064-62-9 | Gd2O3 | 362.50 g/mol
Supplier: Thermo Scientific Chemicals 315510250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysisSpecifications
| Gadolinium(III) oxide | |
| 12064-62-9 | |
| Powder | |
| 99.90% | |
| Gd2O3 | |
| 10, 4200 | |
| Solubility in water: insoluble in water. Other solubilities: soluble in acids | |
| [O--].[O--].[O--].[Gd+3].[Gd+3] | |
| 362.50 | |
| 362.5 | |
| Trace Metal Basis |
| 1900.0°C | |
| White | |
| 25 g | |
| Gd2O3 | |
| MFCD00011026 | |
| gadolinia, gadolinium oxide, gadolinium iii oxide, gadolinium trioxide, digadolinium trioxide, gadolinium sesquioxide, gadolinium 3+ oxide, unii-5480d0nhlj, gadolinium 3+ ; oxygen 2-, digadolinium 3+ ion trioxidandiide | |
| CMIHHWBVHJVIGI-UHFFFAOYSA-N | |
| digadolinium(3+) trioxidandiide | |
| 159427 | |
| 99.9% | |
| Glass bottle |
Chemical Identifiers
| 12064-62-9 | |
| 362.50 | |
| CMIHHWBVHJVIGI-UHFFFAOYSA-N | |
| 159427 |
| Gd2O3 | |
| MFCD00011026 | |
| gadolinia, gadolinium oxide, gadolinium iii oxide, gadolinium trioxide, digadolinium trioxide, gadolinium sesquioxide, gadolinium 3+ oxide, unii-5480d0nhlj, gadolinium 3+ ; oxygen 2-, digadolinium 3+ ion trioxidandiide | |
| [O--].[O--].[O--].[Gd+3].[Gd+3] |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
EINECSNumber : 235-060-9
RUO – Research Use Only