missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) oxide, 99.9%, (trace metal basis)

Catalog No. p-3756641
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC315510100 10 g Glass bottle
AC315510250 25 g Glass bottle
AC315511000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC315510100 Supplier Thermo Scientific Chemicals Supplier No. 315510100
Only null left

CAS: 12064-62-9 | Gd2O3 | 362.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis

Chemical Identifiers

CAS 12064-62-9
Molecular Formula Gd2O3
Molecular Weight (g/mol) 362.50
MDL Number MFCD00011026
InChI Key CMIHHWBVHJVIGI-UHFFFAOYSA-N
Synonym gadolinia, gadolinium oxide, gadolinium iii oxide, gadolinium trioxide, digadolinium trioxide, gadolinium sesquioxide, gadolinium 3+ oxide, unii-5480d0nhlj, gadolinium 3+ ; oxygen 2-, digadolinium 3+ ion trioxidandiide
PubChem CID 159427
SMILES [O--].[O--].[O--].[Gd+3].[Gd+3]

Specifications

Melting Point 1900.0°C
Color White
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.90%
Linear Formula Gd2O3
Merck Index 10, 4200
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in acids
IUPAC Name gadolinium(3+);oxygen(2-)
Formula Weight 362.5
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Gadolinium(III) oxide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 235-060-9

RUO – Research Use Only