Learn More
Blackline Safety™ G7x Satellite-Connected Safety Monitor with Single-Gas Sensor Cartridge, North America
Wireless connectivity ensures that each G7x is remotely configurable and firmware updates are automatic. G7x supports three cartridge types that provide a choice of gas detection options. Everything is completely customizable to meet the needs of every industry, application and scenario
Supplier: Blackline Safety™ G7XSCNA
Description
- Fall detection and No motion detection: Tri-axis accelerometer, tri-axis gyro, software processing, configurable sensitivity, configurable time window (1 to 30 mins, or off ) for no motion detection
- Emergency latch: Pull latch to trigger remote alert
- Silent emergency: Press and hold latch to trigger remote alert
- Low-battery: configurable threshold
- Worker check-in: Configurable timer (5 to 180 min, or off ), automatic check-in when driving
Chemical Identifiers
| Single Gas Cartridge |
Specifications
| Carbon Dioxide | |
| Single Gas Cartridge |