Learn More
Blackline Safety™ G7x Satellite-Connected Safety Monitor with Single-Gas Sensor Cartridge, North America
Catalog No. p-7160370
Change view
Click to view available options
Components Measured:
Ammonia
Carbon Dioxide
Carbon Monoxide
Carbon Monoxide - H
Chlorine
Hydrogen Cyanide
Hydrogen Sulfide
Oxygen
PID
Sulfur Dioxide
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
170000133 Ammonia
170000134 Carbon Dioxide
170000135 Hydrogen Sulfide
170000136 Chlorine
170000137 Carbon Monoxide
170000138 Oxygen
170000139 Carbon Monoxide - H
170000140 Sulfur Dioxide
170000141 PID
170000145 Hydrogen Cyanide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 170000133 Supplier Blackline Safety™ Supplier No. G7XSANA
Wireless connectivity ensures that each G7x is remotely configurable and firmware updates are automatic. G7x supports three cartridge types that provide a choice of gas detection options. Everything is completely customizable to meet the needs of every industry, application and scenario
  • Fall detection and No motion detection: Tri-axis accelerometer, tri-axis gyro, software processing, configurable sensitivity, configurable time window (1 to 30 mins, or off ) for no motion detection
  • Emergency latch: Pull latch to trigger remote alert
  • Silent emergency: Press and hold latch to trigger remote alert
  • Low-battery: configurable threshold
  • Worker check-in: Configurable timer (5 to 180 min, or off ), automatic check-in when driving
Specifications
Components Measured Ammonia
Type Single Gas Cartridge