Learn More
Blackline Safety™ G7c Cellular-Connected Safety Monitor with Multi-Gas Sensor Cartridge, North America
Wireless connectivity ensures that each G7c is remotely configurable and firmware updates are automatic. G7c supports three cartridge types that provide a choice of gas detection options. Everything is completely customizable to meet the needs of every industry, application and scenario
$1,102.70
| Catalog Number | Mfr. No. | Components Measured | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Components Measured | Price | Quantity & Availability | |||||
|
170000002
|
Blackline Safety™
G7CQACIONA |
Methane, Carbon Dioxide, LEL-I, Oxygen |
Each for $1,102.70
|
|
|||||
|
170000004
|
Blackline Safety™
G7CQACOXNA |
Methane, Carbon Dioxide, Oxygen |
Each for $1,102.70
|
|
|||||
|
170000005
|
Blackline Safety™
G7CQACXXNA |
Methane, Carbon Dioxide |
Each for $1,102.70
|
|
|||||
|
170000008
|
Blackline Safety™
G7CQAHIONA |
Methane, Hydrogen Sulfide, LEL-I, Oxygen |
Each for $1,102.70
|
|
|||||
|
170000009
|
Blackline Safety™
G7CQAHOXNA |
Methane, Hydrogen Sulfide, Oxygen |
Each for $1,102.70
|
|
|||||
|
170000010
|
Blackline Safety™
G7CQAILONA |
Methane, LEL-I, Chlorine, Oxygen |
Each for $1,102.70
|
|
|||||
|
170000011
|
Blackline Safety™
G7CQAIMONA |
Methane, LEL-I, Carbon Monoxide, Oxygen |
Each for $1,102.70
|
|
|||||
|
170000012
|
Blackline Safety™
G7CQAIORNA |
Methane, LEL-I, Oxygen, Cobalt Hydride |
Each for $1,102.70
|
|
|||||
|
170000013
|
Blackline Safety™
G7CQAIOSNA |
Methane, LEL-I, Oxygen, Sulfur Dioxide |
Each for $1,102.70
|
|
|||||
|
170000014
|
Blackline Safety™
G7CQAIOXNA |
Methane, LEL-I, Oxygen |
Each for $1,102.70
|
|
|||||
|
170000016
|
Blackline Safety™
G7CQAMXXNA |
Methane, Carbon Monoxide |
Each for $1,102.70
|
|
|||||
|
170000017
|
Blackline Safety™
G7CQAOXXNA |
Methane, Oxygen |
Each for $1,102.70
|
|
|||||
|
170000020
|
Blackline Safety™
G7CQCHIONA |
Carbon Dioxide, Hydrogen Sulfide, LEL-I, Oxygen |
Each for $1,102.70
|
|
|||||
|
170000022
|
Blackline Safety™
G7CQCIOXNA |
Carbon Dioxide, LEL-I, Oxygen |
Each for $1,102.70
|
|
|||||
|
170000025
|
Blackline Safety™
G7CQCOXXNA |
Carbon Dioxide, Oxygen |
Each for $1,102.70
|
|
|||||
|
170000031
|
Blackline Safety™
G7CQHILONA |
Hydrogen Sulfide, LEL-I, Chlorine, Oxygen |
Each for $1,102.70
|
|
|||||
|
170000034
|
Blackline Safety™
G7CQHIOSNA |
Hydrogen Sulfide, LEL-I, Oxygen, Sulfur Dioxide |
Each for $1,102.70
|
|
|||||
|
170000035
|
Blackline Safety™
G7CQHIOVNA |
Hydrogen Sulfide, LEL-I, Oxygen, PID |
Each for $1,102.70
|
|
|||||
|
170000036
|
Blackline Safety™
G7CQHIOXNA |
Hydrogen Sulfide, LEL-I, Oxygen |
Each for $1,102.70
|
|
|||||
|
170000038
|
Blackline Safety™
G7CQHMOXNA |
Hydrogen Sulfide, Carbon Monoxide, Oxygen |
Each for $1,102.70
|
|
|||||
|
170000039
|
Blackline Safety™
G7CQHMXXNA |
Hydrogen Sulfide, Carbon Monoxide |
Each for $1,102.70
|
|
|||||
|
170000042
|
Blackline Safety™
G7CQHORXNA |
Hydrogen Sulfide, Oxygen, Carbon Monoxide - H |
Each for $1,102.70
|
|
|||||
|
170000043
|
Blackline Safety™
G7CQILMONA |
LEL-I, Chlorine, Carbon Monoxide, Oxygen |
Each for $1,102.70
|
|
|||||
|
170000044
|
Blackline Safety™
G7CQILOSNA |
LEL-I, Chlorine, Oxygen, Sulfur Dioxide |
Each for $1,102.70
|
|
|||||
|
170000045
|
Blackline Safety™
G7CQILOXNA |
LEL-I, Chlorine, Oxygen |
Each for $1,102.70
|
|
|||||
|
170000046
|
Blackline Safety™
G7CQIMOSNA |
LEL-I, Carbon Monoxide, Oxygen, Sulfur Dioxide |
Each for $1,102.70
|
|
|||||
|
170000047
|
Blackline Safety™
G7CQIMOXNA |
LEL-I, Carbon Monoxide, Oxygen |
Each for $1,102.70
|
|
|||||
|
170000048
|
Blackline Safety™
G7CQIOVXNA |
LEL-I, Oxygen, PID |
Each for $1,102.70
|
|
|||||
|
170000049
|
Blackline Safety™
G7CQIOXXNA |
LEL-I, Oxygen |
Each for $1,102.70
|
|
|||||
|
170000050
|
Blackline Safety™
G7CQIVXXNA |
LEL-I, PID |
Each for $1,102.70
|
|
|||||
|
170000055
|
Blackline Safety™
G7CQMOXXNA |
Carbon Monoxide, Oxygen |
Each for $1,102.70
|
|
|||||
|
170000059
|
Blackline Safety™
G7CQOVXXNA |
Oxygen, PID |
Each for $1,102.70
|
|
|||||
|
170000023
|
Blackline Safety™
G7CQCMOXNA |
Carbon Dioxide, Oxygen |
Each for $1,102.70
|
|
|||||
Description
- Fall detection and No motion detection: Tri-axis accelerometer, tri-axis gyro, software processing, configurable sensitivity, configurable time window (1 to 30 mins, or off ) for no motion detection
- Emergency latch: Pull latch to trigger remote alert
- Silent emergency: Press and hold latch to trigger remote alert
- Low-battery: configurable threshold
- Worker check-in: Configurable timer (5 to180 min, or off ), automatic check-in when driving