Learn More
- Home
- Products
- Industrial Hygiene and Environmental Monitoring
- Gas and Vapor Detection
- Multigas Portable Detectors
- Blackline Safety™ G7c Cellular-Connected Safety Monitor with Multi-Gas Sensor Cartridge, North America
Blackline Safety™ G7c Cellular-Connected Safety Monitor with Multi-Gas Sensor Cartridge, North America
Catalog No. 170000044
Change view
Click to view available options
Components Measured:
Carbon Dioxide, Hydrogen Sulfide, LEL-I, Oxygen
Carbon Dioxide, LEL-I, Oxygen
Carbon Dioxide, Oxygen
Carbon Monoxide, Oxygen
Hydrogen Sulfide, Carbon Monoxide
Hydrogen Sulfide, Carbon Monoxide, Oxygen
Hydrogen Sulfide, LEL-I, Chlorine, Oxygen
Hydrogen Sulfide, LEL-I, Oxygen
Hydrogen Sulfide, LEL-I, Oxygen, PID
Hydrogen Sulfide, LEL-I, Oxygen, Sulfur Dioxide
Hydrogen Sulfide, Oxygen, Carbon Monoxide - H
LEL-I, Carbon Monoxide, Oxygen
LEL-I, Carbon Monoxide, Oxygen, Sulfur Dioxide
LEL-I, Chlorine, Carbon Monoxide, Oxygen
LEL-I, Chlorine, Oxygen
LEL-I, Chlorine, Oxygen, Sulfur Dioxide
LEL-I, Oxygen
LEL-I, Oxygen, PID
LEL-I, PID
Methane, Carbon Dioxide
Methane, Carbon Dioxide, LEL-I, Oxygen
Methane, Carbon Dioxide, Oxygen
Methane, Carbon Monoxide
Methane, Hydrogen Sulfide, LEL-I, Oxygen
Methane, Hydrogen Sulfide, Oxygen
Methane, LEL-I, Carbon Monoxide, Oxygen
Methane, LEL-I, Chlorine, Oxygen
Methane, LEL-I, Oxygen
Methane, LEL-I, Oxygen, Cobalt Hydride
Methane, LEL-I, Oxygen, Sulfur Dioxide
Methane, Oxygen
Oxygen, PID
33 product options available for selection
| Catalog No. | Components Measured |
|---|---|
| 170000044 | LEL-I, Chlorine, Oxygen, Sulfur Dioxide |
| 170000002 | Methane, Carbon Dioxide, LEL-I, Oxygen |
| 170000004 | Methane, Carbon Dioxide, Oxygen |
| 170000005 | Methane, Carbon Dioxide |
| 170000008 | Methane, Hydrogen Sulfide, LEL-I, Oxygen |
| 170000009 | Methane, Hydrogen Sulfide, Oxygen |
| 170000010 | Methane, LEL-I, Chlorine, Oxygen |
| 170000011 | Methane, LEL-I, Carbon Monoxide, Oxygen |
| 170000012 | Methane, LEL-I, Oxygen, Cobalt Hydride |
| 170000013 | Methane, LEL-I, Oxygen, Sulfur Dioxide |
| 170000014 | Methane, LEL-I, Oxygen |
| 170000016 | Methane, Carbon Monoxide |
| 170000017 | Methane, Oxygen |
| 170000020 | Carbon Dioxide, Hydrogen Sulfide, LEL-I, Oxygen |
| 170000022 | Carbon Dioxide, LEL-I, Oxygen |
| 170000025 | Carbon Dioxide, Oxygen |
| 170000031 | Hydrogen Sulfide, LEL-I, Chlorine, Oxygen |
| 170000034 | Hydrogen Sulfide, LEL-I, Oxygen, Sulfur Dioxide |
| 170000035 | Hydrogen Sulfide, LEL-I, Oxygen, PID |
| 170000036 | Hydrogen Sulfide, LEL-I, Oxygen |
| 170000038 | Hydrogen Sulfide, Carbon Monoxide, Oxygen |
| 170000039 | Hydrogen Sulfide, Carbon Monoxide |
| 170000042 | Hydrogen Sulfide, Oxygen, Carbon Monoxide - H |
| 170000043 | LEL-I, Chlorine, Carbon Monoxide, Oxygen |
| 170000045 | LEL-I, Chlorine, Oxygen |
| 170000046 | LEL-I, Carbon Monoxide, Oxygen, Sulfur Dioxide |
| 170000047 | LEL-I, Carbon Monoxide, Oxygen |
| 170000048 | LEL-I, Oxygen, PID |
| 170000049 | LEL-I, Oxygen |
| 170000050 | LEL-I, PID |
| 170000055 | Carbon Monoxide, Oxygen |
| 170000059 | Oxygen, PID |
| 170000023 | Carbon Dioxide, Oxygen |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 33 options available.
Description
- Fall detection and No motion detection: Tri-axis accelerometer, tri-axis gyro, software processing, configurable sensitivity, configurable time window (1 to 30 mins, or off ) for no motion detection
- Emergency latch: Pull latch to trigger remote alert
- Silent emergency: Press and hold latch to trigger remote alert
- Low-battery: configurable threshold
- Worker check-in: Configurable timer (5 to180 min, or off ), automatic check-in when driving
Specifications
Specifications
| Components Measured | LEL-I, Chlorine, Oxygen, Sulfur Dioxide |
| Type | Multigas Cartridge |