missing translation for 'onlineSavingsMsg'
Learn More

Furosemide, 97+%

Catalog No. AAJ6145706
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6145706 5 g
AAJ6145709 10 g
AAJ6145714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6145706 Supplier Thermo Scientific Chemicals Supplier No. J6145706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Loop diuretic that inhibits the Na+/2Cl-/K+ (NKCC) cotransporter | CAS: 54-31-9 | C12H11ClN2O5S | 330.739 g/mol

Loop diuretic that inhibits the Na+/2Cl-/K+ (NKCC) cotransporter

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54-31-9
Molecular Formula C12H11ClN2O5S
Molecular Weight (g/mol) 330.739
MDL Number MFCD00010549
InChI Key ZZUFCTLCJUWOSV-UHFFFAOYSA-N
Synonym furosemide, frusemide, lasix, furosemid, furanthril, errolon, fusid, aisemide, beronald, desdemin
PubChem CID 3440
ChEBI CHEBI:47426
IUPAC Name 4-chloro-2-(furan-2-ylmethylamino)-5-sulfamoylbenzoic acid
SMILES C1=COC(=C1)CNC2=CC(=C(C=C2C(=O)O)S(=O)(=O)N)Cl

Specifications

Melting Point 206°C to 209°C
Color White
Quantity 5 g
Merck Index 14,4309
Solubility Information Soluble in acetone, DMF or methanol; Slightly soluble in water
Formula Weight 330.74
Percent Purity ≥97%
Physical Form Crystalline Powder
Chemical Name or Material Furosemide
Hazard Category H360
Hazard Statement GHS H Statement
H360
May damage fertility or the unborn child.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 200-203-6
RTECSNumber CB2625000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only