missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fumaric Acid 99.0+%, TCI America™
$37.97 - $47.46
Chemical Identifiers
| CAS | 110-17-8 |
|---|---|
| Molecular Formula | C4H4O4 |
| Molecular Weight (g/mol) | 116.07 |
| MDL Number | MFCD00002700 |
| InChI Key | VZCYOOQTPOCHFL-OWOJBTEDSA-N |
| Synonym | fumaric acid, 2-butenedioic acid, trans-butenedioic acid, allomaleic acid, boletic acid, lichenic acid, tumaric acid, 2e-but-2-enedioic acid, butenedioic acid, trans-2-butenedioic acid |
| PubChem CID | 444972 |
| ChEBI | CHEBI:18012 |
| IUPAC Name | (2E)-but-2-enedioic acid |
| SMILES | OC(=O)\C=C\C(O)=O |
Chemical Identifiers
| C4H4O4 | |
| MFCD00002700 | |
| fumaric acid, 2-butenedioic acid, trans-butenedioic acid, allomaleic acid, boletic acid, lichenic acid, tumaric acid, 2e-but-2-enedioic acid, butenedioic acid, trans-2-butenedioic acid | |
| CHEBI:18012 | |
| OC(=O)\C=C\C(O)=O |
Specifications
| 110-17-8 | |
| White | |
| C4H4O4 | |
| fumaric acid, 2-butenedioic acid, trans-butenedioic acid, allomaleic acid, boletic acid, lichenic acid, tumaric acid, 2e-but-2-enedioic acid, butenedioic acid, trans-2-butenedioic acid | |
| OC(=O)\C=C\C(O)=O | |
| 116.07 | |
| CHEBI:18012 | |
| ≥99.0% (GC,T) | |
| Fumaric Acid |
| 287°C | |
| 2.2 to 2.6 (1% H2O suspention) | |
| MFCD00002700 | |
| VZCYOOQTPOCHFL-OWOJBTEDSA-N | |
| (2E)-but-2-enedioic acid | |
| 444972 | |
| 116.07 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-1091
RTECSNumber : LS9625000
TSCA : Yes