missing translation for 'onlineSavingsMsg'
Learn More

Fumaric Acid, NF, 99.5-100.5%, Spectrum™ Chemical

F1096, 110-17-8, C4H4O4

$354.00 - $15,499.91

Chemical Identifiers

CAS 110-17-8
Molecular Formula C4H4O4
Molecular Weight (g/mol) 116.07
MDL Number MFCD00002700
InChI Key VZCYOOQTPOCHFL-OWOJBTEDSA-N
IUPAC Name (2E)-but-2-enedioic acid
SMILES OC(=O)\C=C\C(O)=O
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18602874
View Documents Promotion Details
N/A 100 g Poly Bottle
Each for $354.00
Only null left
 
18602870
View Documents Promotion Details
N/A 500 g Poly Bottle
Each for $649.39
Only null left
 
18602871
View Documents Promotion Details
N/A 2.5 kg Poly Bottle
Each for $1,862.42
Only null left
 
18602872
View Documents Promotion Details
N/A 12 kg Poly Bottle
Each for $6,113.97
Only null left
 
18602873
View Documents Promotion Details
N/A 50 kg Poly Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Fumaric Acid, NF is used an effervescent agent in tablet formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

110-17-8
116.07
VZCYOOQTPOCHFL-OWOJBTEDSA-N
OC(=O)\C=C\C(O)=O
C4H4O4
MFCD00002700
(2E)-but-2-enedioic acid

Specifications

110-17-8
290°C
C4H4O4
VZCYOOQTPOCHFL-OWOJBTEDSA-N
(2E)-but-2-enedioic acid
99.5 to 100.5%
Crystalline Powder or Lumps
100%
0.1%
MFCD00002700
OC(=O)\C=C\C(O)=O
116.07
NF
Fumaric Acid
Videos
SDS
Documents

Documents