missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fumaric Acid, NF, 99.5-100.5%, Spectrum™ Chemical
F1096, 110-17-8, C4H4O4
Supplier: Spectrum Chemical Mfg Cor F10962.5KG
| Quantity | 2.5 kg |
|---|---|
| Packaging | Poly Bottle |
Description
Spectrum™ Chemical Fumaric Acid, NF is used an effervescent agent in tablet formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 110-17-8 | |
| 116.07 | |
| VZCYOOQTPOCHFL-OWOJBTEDSA-N | |
| OC(=O)\C=C\C(O)=O |
| C4H4O4 | |
| MFCD00002700 | |
| (2E)-but-2-enedioic acid |
Specifications
| Fumaric Acid | |
| 100% | |
| 0.1% | |
| C4H4O4 | |
| 2.5 kg | |
| OC(=O)\C=C\C(O)=O | |
| 116.07 | |
| NF |
| 110-17-8 | |
| 290°C | |
| Poly Bottle | |
| MFCD00002700 | |
| VZCYOOQTPOCHFL-OWOJBTEDSA-N | |
| (2E)-but-2-enedioic acid | |
| 99.5 to 100.5% | |
| Crystalline Powder or Lumps |