missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fulvestrant

Catalog No. AC465782500
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465782500 250 mg
AC465780010 1 g
AC465780500 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC465782500 Supplier Thermo Scientific Chemicals Supplier No. 465782500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 129453-61-8
Molecular Formula C32 H47 F5 O3 S

Specifications

Quantity 250 mg
Solubility (0.1 g in 10 mL Ethanol) Clear solution
Formula Weight 606.77
Physical Form Powder
Product Type Reagent
Purity ≥98.5% (HPLC)
Chemical Name or Material Fulvestrant

RUO – Research Use Only