missing translation for 'onlineSavingsMsg'
Learn More

Fullerene powder, 99+% C{70}

CAS: 115383-22-7 | C70 | 840.77 g/mol

Supplier:  Thermo Scientific Chemicals 04260003

 View more versions of this product

Catalog No. AA4260003

Only null left
Explore available promotions

Description

Description

Fullerene powder is used for biomedical applications including the design of high-performance magnetic resonance imaging (MRI) contrast agents, X-ray imaging contrast agents, photodynamic therapy and drug and gene delivery. It is used in superconductors, photoactive polymers, film material modifiers, abrasive agent for cutting and grinding applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Fullerene powder
Gray to Black
500°C to 600°C (sublimation)
C70
1 g
fullerene, fullerene 70, rugbyballene, carbon, 70-d5h fullerene, buckminsterfullerene, 5,6 fullerene-c70-d5h 6, c70-d5h 6 5,6 fullerene, fullerene powder, c 250mg
ATLMFJTZZPOKLC-UHFFFAOYSA-N
(C\{70}-D\{5h(6)})[5,6]fullerene
16131935
≥99%
115383-22-7
1.7 g/mL
C70
MFCD00282904
UN1325
Soluble in benzene and toluene. Insoluble in water.
C12=C3C4=C5C6=C7C8=C9C%10=C%11C%12=C%13C%10=C%10C8=C5C1=C%10C1=C%13C5=C8C1=C2C1=C3C2=C3C%10=C%13C%14=C3C1=C8C1=C3C5=C%12C5=C8C%11=C%11C9=C7C7=C9C6=C4C2=C2C%10=C4C(=C29)C2=C6C(=C8C8=C9C6=C4C%13=C9C(=C%141)C3=C85)C%11=C27
840.77
CHEBI:33195
Powder

Chemical Identifiers

115383-22-7
840.77
ATLMFJTZZPOKLC-UHFFFAOYSA-N
16131935
(C\{70}-D\{5h(6)})[5,6]fullerene
C70
MFCD00282904
fullerene, fullerene 70, rugbyballene, carbon, 70-d5h fullerene, buckminsterfullerene, 5,6 fullerene-c70-d5h 6, c70-d5h 6 5,6 fullerene, fullerene powder, c 250mg
CHEBI:33195
C12=C3C4=C5C6=C7C8=C9C%10=C%11C%12=C%13C%10=C%10C8=C5C1=C%10C1=C%13C5=C8C1=C2C1=C3C2=C3C%10=C%13C%14=C3C1=C8C1=C3C5=C%12C5=C8C%11=C%11C9=C7C7=C9C6=C4C2=C2C%10=C4C(=C29)C2=C6C(=C8C8=C9C6=C4C%13=C9C(=C%141)C3=C85)C%11=C27
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H228-H315-H319-H335

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only