missing translation for 'onlineSavingsMsg'
Learn More

Fullerene powder, 98+% C{70}

Catalog No. p-7052126
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42601MD 250 mg
AA4260103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42601MD Supplier Thermo Scientific Chemicals Supplier No. 042601MD
Only null left

CAS: 115383-22-7 | C70 | 840.77 g/mol

Fullerene powder is used in chemical research. It is extensively used for several biomedical applications including the design of high-performance MRI contrast agents, X-Ray imaging contrast agents, photodynamic therapy and drug and gene delivery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 115383-22-7
Molecular Formula C70
Molecular Weight (g/mol) 840.77
MDL Number MFCD00282904
InChI Key ATLMFJTZZPOKLC-UHFFFAOYSA-N
Synonym fullerene, fullerene 70, rugbyballene, carbon, 70-d5h fullerene, buckminsterfullerene, 5,6 fullerene-c70-d5h 6, c70-d5h 6 5,6 fullerene, fullerene powder, c 250mg
PubChem CID 16131935
ChEBI CHEBI:33195
IUPAC Name (C\{70}-D\{5h(6)})[5,6]fullerene
SMILES C12=C3C4=C5C6=C7C8=C9C%10=C%11C%12=C%13C%10=C%10C8=C5C1=C%10C1=C%13C5=C8C1=C2C1=C3C2=C3C%10=C%13C%14=C3C1=C8C1=C3C5=C%12C5=C8C%11=C%11C9=C7C7=C9C6=C4C2=C2C%10=C4C(=C29)C2=C6C(=C8C8=C9C6=C4C%13=C9C(=C%141)C3=C85)C%11=C27

Specifications

Color Black
Density ≈1.7 g/mL
Boiling Point 500°C to 600°C (sublimation)
Quantity 250 mg
UN Number UN1325
Solubility Information Soluble in toluene and benzene.
Percent Purity ≥98%
Physical Form Powder
Assay C70
Chemical Name or Material Fullerene powder
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only