missing translation for 'onlineSavingsMsg'
Learn More

Fullerene C70 98.0+%, TCI America™

Catalog No. B1694100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1694100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B1694100MG Supplier TCI America Supplier No. B1694100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 115383-22-7
Molecular Formula C70
Molecular Weight (g/mol) 840.77
MDL Number MFCD00146976
InChI Key ATLMFJTZZPOKLC-UHFFFAOYSA-N
Synonym fullerene, fullerene 70, rugbyballene, carbon, 70-d5h fullerene, buckminsterfullerene, 5,6 fullerene-c70-d5h 6, c70-d5h 6 5,6 fullerene, fullerene powder, c 250mg
PubChem CID 16131935
ChEBI CHEBI:33195
IUPAC Name (C\{70}-D\{5h(6)})[5,6]fullerene
SMILES C12=C3C4=C5C6=C7C8=C9C%10=C%11C%12=C%13C%10=C%10C8=C5C1=C%10C1=C%13C5=C8C1=C2C1=C3C2=C3C%10=C%13C%14=C3C1=C8C1=C3C5=C%12C5=C8C%11=C%11C9=C7C7=C9C6=C4C2=C2C%10=C4C(=C29)C2=C6C(=C8C8=C9C6=C4C%13=C9C(=C%141)C3=C85)C%11=C27

Specifications

Color Black-Green
Quantity 100 mg
Formula Weight 840.77
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Fullerene C70