missing translation for 'onlineSavingsMsg'
Learn More

Fructose Standard, 30,000 ppm C6H12O6, Ricca Chemical

Catalog No. R3219030100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R3219030100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R3219030100 Supplier Ricca Chemical Company Supplier No. R3219030100A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Subject to microbiological contamination. Refrigeration may extend shelf life.

Chemical Identifiers

CAS 57-48-7 , 7732-18-5
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
InChI Key BJHIKXHVCXFQLS-UYFOZJQFSA-N
PubChem CID 5984
ChEBI CHEBI:48095
IUPAC Name (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES C(C(C(C(C(=O)CO)O)O)O)O

Specifications

Packaging Natural Poly Bottle
Odor Odorless
Physical Form Liquid
Color Colorless to Yellow
pH Neutral
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Solution
Type Single-component Standard
Quantity 100 mL
Grade Laboratory
Chemical Name or Material Fructose Standard, 30, 000 ppm
Show More Show Less
Recommended Storage 2°C - 8°C (36°F - 46°F)