missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hughes Freeze Protected, Floor Mount Combination Shower with Closed ABS Bowl

Hughes Freeze Protected, Floor Mount Combination Shower is intended for use where there is a possibility of the water inside of the shower freezing due to extreme cold climates
$7,019.48 - $9,395.72
Specifications
| Activation | Pull Handle, Pull Lid |
|---|---|
| Bowl Size (Metric) | 241 mm |
| Certifications/Compliance | ANSI Z358.1-2014 and EN15154 Compliant |
| Color | White with Green Stripes |
| Depth (English) | 24 in. |
| Catalog Number | Mfr. No. | Electrical Requirements | Material (Piping Connection) | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Electrical Requirements | Material (Piping Connection) | Price | Quantity & Availability | ||||
18666699
![]() |
N/A | 120 V | Galvanized Steel |
Each for $7,019.48
|
|
||||
18666700
![]() |
N/A | 120 V | Stainless Steel |
Each for $9,395.72
|
|
||||
18666701
![]() |
N/A | 240 V | Galvanized Steel |
Each for $7,019.48
|
|
||||
18666702
![]() |
N/A | 240 V | Stainless Steel |
Each for $9,395.72
|
|
||||
Description
Features:
- Trace tape heating with polyethylene jacket and polyurethane foam insulation to improve frost protection and strength.
- Specially designed, high-performance quarter-turn ball valve made of 316L stainless steel to endure the harshest environments and site conditions.
- Low operating torque to open valve offers reliable, easy on/off operation when emergencies occur.
- Valve includes an integral, heavy-duty stainless steel stop-plate to handle rough operation without failure.
- Durable, coated stainless steel shower pull handle with nylon valve connection is easy to grip and activate for immediate water drenching.
- Includes a removable, self-draining overhead nozzle designed for high flow rates that meet or exceed ANSI standards.
- Constructed from a durable nylon material, the nozzle offers optimal spray distribution for user protection.
- Made of tough, corrosion-resistant acrylic capped ABS, the covered eye/face wash incorporates an integral lid connected to the water valve for automatic operation—activate by simply pulling down the lid.
- Easy-to-reach flow regulator valve positioned on side of bowl adjusts to suit the severity of the accident.
- Includes an inspection record tag for recording service and test history.
- For cold climates with a minimum ambient operating temperature of -4°F (-20°C) or above
- Covered acrylic-capped ABS eye/face wash protects the bowl and eyewash diffusers from dust and debris
- Suitable for both Class 1 Division 2 hazardous areas, as well as non-hazardous areas
- Includes a universal safety shower sign for enhanced visibility
- Meets ANSI Z358.1 2014 and EN 15154 standards, supports OSHA requirements
Compliance
ANSI Z358.1-2014 and EN15154 Compliant
Warranty
1 Year
Specifications
| Pull Handle, Pull Lid | |
| ANSI Z358.1-2014 and EN15154 Compliant | |
| 24 in. | |
| 3 gpm | |
| 48 in. | |
| 1 Unit | |
| Polyurethane Foam Insulation, Impact Resistant Medium Density Polyethylene, Nylon 6 | |
| 29 to 87 psi | |
| 24 in. |
| 241 mm | |
| White with Green Stripes | |
| 610 mm | |
| Emergency Shower and Eye Wash with Water | |
| 1219 mm | |
| 1.25 in. (F) NPT | |
| Floor | |
| Safety Shower and Emergency Eye Wash | |
| 610 mm |