missing translation for 'onlineSavingsMsg'
Learn More

Hughes Freeze Protected, Floor Mount Combination Shower with Closed ABS Bowl
GREENER_CHOICE

Catalog No. 18666699
Change view
Click to view available options
Electrical Requirements:
120 V
240 V
Material (Piping Connection):
Galvanized Steel
Stainless Steel
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Electrical Requirements Material (Piping Connection)
18666699 120 V Galvanized Steel
18666700 120 V Stainless Steel
18666701 240 V Galvanized Steel
18666702 240 V Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18666699 Supplier Justrite International Supplier No. H5G45G1H

Hughes Freeze Protected, Floor Mount Combination Shower is intended for use where there is a possibility of the water inside of the shower freezing due to extreme cold climates

Features:

  • Trace tape heating with polyethylene jacket and polyurethane foam insulation to improve frost protection and strength.
  • Specially designed, high-performance quarter-turn ball valve made of 316L stainless steel to endure the harshest environments and site conditions.
  • Low operating torque to open valve offers reliable, easy on/off operation when emergencies occur.
  • Valve includes an integral, heavy-duty stainless steel stop-plate to handle rough operation without failure.
  • Durable, coated stainless steel shower pull handle with nylon valve connection is easy to grip and activate for immediate water drenching.
  • Includes a removable, self-draining overhead nozzle designed for high flow rates that meet or exceed ANSI standards.
  • Constructed from a durable nylon material, the nozzle offers optimal spray distribution for user protection.
  • Made of tough, corrosion-resistant acrylic capped ABS, the covered eye/face wash incorporates an integral lid connected to the water valve for automatic operation—activate by simply pulling down the lid.
  • Easy-to-reach flow regulator valve positioned on side of bowl adjusts to suit the severity of the accident.
  • Includes an inspection record tag for recording service and test history.
  • For cold climates with a minimum ambient operating temperature of -4°F (-20°C) or above
  • Covered acrylic-capped ABS eye/face wash protects the bowl and eyewash diffusers from dust and debris
  • Suitable for both Class 1 Division 2 hazardous areas, as well as non-hazardous areas
  • Includes a universal safety shower sign for enhanced visibility
  • Meets ANSI Z358.1 2014 and EN 15154 standards, supports OSHA requirements

Compliance

ANSI Z358.1-2014 and EN15154 Compliant

Warranty

1 Year

Specifications

Activation Pull Handle, Pull Lid
Bowl Size (Metric) 241 mm
Certifications/Compliance ANSI Z358.1-2014 and EN15154 Compliant
Color White with Green Stripes
Depth (English) 24 in.
Depth (Metric) 610 mm
Electrical Requirements 120 V
Flow Rate 3 gpm
For Use With (Application) Emergency Shower and Eye Wash with Water
Height (English) 48 in.
Height (Metric) 1219 mm
Includes 1 Unit
Inlet Connection (Metric) 1.25 in. (F) NPT
Material Polyurethane Foam Insulation, Impact Resistant Medium Density Polyethylene, Nylon 6
Material (Piping Connection) Galvanized Steel
Mounting Floor
Pressure 29 to 87 psi
Product Type Safety Shower and Emergency Eye Wash
Width (English) 24 in.
Width (Metric) 610 mm
Show More Show Less