missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ FreeStyle™ MAX Reagent

Animal origin-free formulation for transfecting plasmid DNA into eukaryotic cells that can easily be scaled up to produce large amounts of recombinant proteins

$1,192.00 - $14,180.00

Specifications

Classification Animal Origin-Free
Cell Type Established Cell Lines
Sample Type Plasmid DNA
Transfection Technique Lipid-based Transfection
High-throughput Compatibility Not High-throughput Compatible (Manual)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
16447100
Encompass
View Documents Promotion Details
Gibco™
16447100
1 mL
Each for $1,192.00
Only null left
 
16447500
Encompass
View Documents Promotion Details
Gibco™
16447500
15 mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

  • Allows highest expression levels and transfection rates with lowest cytotoxicity in bio-production applications
  • Formulated for use with:
  • FreeStyle 293-F Cells (suspension human embryonal kidney cells, Cat. No. R790-07) in serum-free FreeStyle 293 Expression Medium (Cat. No. 12338-018)
  • FreeStyle CHO-S Cells (suspension Chinese Hamster Ovary cells, Cat. No. R800-07) in serum-free FreeStyle CHO Expression Medium (Cat. No. 12651-014)
  • DG44 Cells (DHFR– suspension CHO cells) in CD DG44 Medium with 8 mM L-glutamine and 18 mL/L 10% Pluronic F-68 (Cat. No. A11000-01)
  • FreeStyle™ MAX Reagent is a proprietary, animal origin–free formulation for transfecting plasmid DNA into eukaryotic cells that can easily be scaled up to produce large amounts of recombinant proteins
  • Intended for use with the FreeStyle™ MAX 293 Expression System (Cat. No. K9000-10) and the FreeStyle™ MAX CHO Expression System (Cat. No. K9000-20).
Specifications

Specifications

Animal Origin-Free
Plasmid DNA
Not High-throughput Compatible (Manual)
Transfection Reagent
Wet Ice
Established Cell Lines
Lipid-based Transfection
Freestyle
Yes
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.