missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Forskolin
Description
TB1099-RMU is an Ancillary Material Grade version of Forskolin (Cat. No. 1099). Suitable for use as an ancillary reagent in the further manufacturing of cell therapies.
Specifications
Specifications
| Chemical Name or Material | (3 R,4 aR,5 S,6 S,6 AS,10 S,10 aR,10 bS)-5-(Acetyloxy)-3-ethenyldodecahydro-6,10,10 b-trihydroxy-3,4 A,7,7,10 A-pentamethyl-1 H-naphtho[2,1-b]pyran-1-one |
| CAS | 66575-29-9 |
| Quantity | 10 mg |
| Product Line | Tocris |
| Target | Adenylyl Cyclase Activators |
| Molecular Formula | C22H34O7 |
| Purity | 0.98 |