missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formic Acid, 2% w/v, Reagents
Formic Acid, 2% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.
Supplier: Labchem Inc CF14730020F
Specifications
| Formic Acid, 2% w/v | |
| 7732-18-5,64-18-6 | |
| Colorless | |
| Methanoic Acid, Formylic Acid, Aminic Acid | |
| O | |
| 18.02 | |
| Reagent | |
| Cubitainer | |
| 1.0047 |
| 2 | |
| 100°C | |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| 2% | |
| 20 L | |
| Liquid |
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Methanoic Acid, Formylic Acid, Aminic Acid | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |