missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid, 2% w/v, Reagents

Catalog No. p-200021473
Change view
Click to view available options
Quantity:
2.5 L
10 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335187 10 L
01335186 2.5 L
01335185 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01335187 Supplier Labchem Inc Supplier No. CF14730010F
Only null left

Formic Acid, 2% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 64-18-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Methanoic Acid, Formylic Acid, Aminic Acid
IUPAC Name water
SMILES O

Specifications

pH 2
Boiling Point 100°C
Color Colorless
Percent Purity 2%
Grade Reagent
Quantity 10 L
Packaging Cubitainer
Physical Form Liquid
Specific Gravity 1.0047
Chemical Name or Material Formic Acid, 2% w/v