missing translation for 'onlineSavingsMsg'
Learn More

Formalin Solution, 10% (Histological), Fisher Chemical™

Catalog No. p-22247
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Poly Bottle
PolyPac™
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SF984 4 L Poly Bottle
SF9820 20 L PolyPac™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SF984 Supplier Thermo Fisher Scientific Supplier No. SF98 4
Only null left

Chemical Identifiers

CAS 50-00-0 , 67-56-1 , 7732-18-5
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Physical Form Liquid
Melting Point 0°C
Boiling Point 100°C
Packaging Poly Bottle
Color Colorless
Quantity 4 L
Percent Purity 3.5 to 4.5%
Grade Histological
Concentration or Composition (by Analyte or Components) 10% w/v
Chemical Name or Material Formalin Solution, 10%
Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Cancer hazard. Cannot be made non-poisonous. Harmful if absorbed through skin or if inhaled. Vapor harmful. Irritating to eyes, respiratory system and skin. May cause an allergic skin reaction. Possible risks of irreversible effects. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:2
Flammability:1
Instability:0
DOTInformation DOT Class 9, : Miscellaneous