missing translation for 'onlineSavingsMsg'
Learn More

Formalin, Buffered, 10% (Acetate Buffer/Certified), Fisher Chemical™

Supplier:  Thermo Fisher Scientific SF99 20

 View more versions of this product

Catalog No. SF9920


Get Up to $1,800 in Chemicals with Purchase
View all promotions
Only null left

Description

Description

Specifications

Chemical Identifiers

Liquid
50-00-0 , 67-56-1 , 127-09-3 , 7732-18-5
7
4.0 to 5.0%
10% w/v (Acetate Buffer)
0°C
1.10g/cm³
MFCD00003274
Certified
Formalin, Buffered, 10%, Acetate Buffer

Specifications

Liquid
0°C
50-00-0 , 67-56-1 , 127-09-3 , 7732-18-5
PolyPac™
20 L
4.0 to 5.0%
10% w/v (Acetate Buffer)
Formalin, Buffered, 10%
Acetate Buffer
1.10g/cm³
7
MFCD00003274
Certified
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Cancer hazard. May cause an allergic skin reaction. Irritating to eyes, respiratory system and skin. May be harmful by inhalation, in contact with skin and if swallowed. May cause central nervous system effects. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:2
Instability:0

DOTInformation : DOT Class 9, : Miscellaneous

SDS
Documents

Documents

Promotions

Promotions