missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde Soln., 4%, Buffered, pH 6.9 (approx. 10% Formalin soln.), For Histology, MilliporeSigma™

Supplier:  Technidata 1004969010

 View more versions of this product

Catalog No. M1004969010


Only null left
Explore available promotions

Description

Description

For histology
Specifications

Specifications

Liquid
approx. 10% Formalin solution
Characteristic odor
Colorless
4%
Formaldehyde solution
50-00-0,67-56-1
Plastic container
10 L
Formaline solution, Methanal solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions