missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formaldehyde - Sodium Carbonate Solution, Ricca Chemical
Specifications
Specifications
| CAS Min % | 41.89, 56.0, 0.33 |
| CAS Max % | 43.6, 58.0, 0.35 |
| Color | Clear colorless |
| Boiling Point | Approximately 100°C |
| Packaging | Amber Poly Bottle |
| Quantity | 500 mL |
| Grade | Laboratory |
| Physical Form | Liquid |
| Chemical Name or Material | Formaldehyde - Sodium Carbonate Solution |
Safety and Handling
| ShelfLife | 24 months |