missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde, Purified. 37%, Reagents

Catalog No. 01337238
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337238 1 L
01337237 -
01337236 4 L
01337235 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01337238 Supplier Labchem Inc Supplier No. C22085001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Formaldehyde, Purified. 37%, Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 50-00-0 , 67-56-1
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Formalin, Methanal
IUPAC Name water
SMILES O

Specifications

Boiling Range 96°C
Color Colorless
Boiling Point 96°C
Packaging Poly Bottle
Quantity 1 L
Specific Gravity 1.0975
Grade Technical
Physical Form Liquid
Chemical Name or Material Formaldehyde, Purified. 37%