missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Formaldehyde, ACS reagent, 37 wt% sol., stab. 10-15% methanol

Formaldehyde, formalin, CAS # 50-00-0, is a colorless volatile used as a histologic fixative and disinfectant. | CAS: 50-00-0 | CH2O | 30.03 g/mol

Supplier:  Thermo Scientific Chemicals 410731000

 View more versions of this product

Catalog No. AC410731000


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Formaldehyde (formalin) is an organic compound used for crosslinking proteins in biological samples such as tissue sections, culture cells, and whole specimens
  • Formaldehyde is an intermediate in the combustion of methane and can exist in many forms
  • It is a volatile, colorless liquid; can be stored long-term in appropriate containers
  • Comes stabilized in solution with methanol

Applications

  • May be used in pain research for induction of pain and inflammation
  • Monomeric formaldehyde is formulated especially for laboratory applications
  • Commonly used in RNA isolation as a denaturant
Specifications

Specifications

Formaldehyde
50-00-0 , 7732-18-5 , 67-56-1
Clear
97.0°C
Plastic bottle
1.3755 to 1.3775
MFCD00003274
01,397; 02,200; 04,238; 05,312; 06,264; 07,158; 08,231; 09,224; 10,186; 11,240; 12,232; 14,168; 15,161; 16,171
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
WSFSSNUMVMOOMR-UHFFFAOYSA-N
formaldehyde
10 to 15% methanol
CHEBI:16842
2mbar at 20°C
ACS Reagent
37 wt% Solution
-15.0°C
1.0830g/mL
50°C
CH2O
HCHO
100 g
15, 4263
Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and acetone
C=O
30.03
712
30.03
36.5 to 38.0%
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only