missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde, 37%, Solution, Spectrum™ Chemical

Catalog No. p-7157457
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
Packaging:
Drum
Poly Bottle
Poly Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602830 500 mL Poly Bottle
18602829 1 L Poly Bottle
18602826 4 L Poly Bottle
18602827 20 L Poly Drum
18602828 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18602830 Supplier Spectrum Chemical Mfg Cor Supplier No. F3328500ML
Only null left

F3328, 50-00-0, HCHO

Spectrum™ Chemical Formaldehyde, 37 Percent, Solution is also known as methanal and is the simplest aldehyde. It has many uses, one of which is formol, the famous embalming and tissue preserving liquid. It is also used in industrial application for the production of different types of resins. It is also a well known tissue fixative and was formerly a disinfectant.

Chemical Identifiers

CAS 50-00-0 , 7732-18-5
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
IUPAC Name formaldehyde
SMILES C=O

Specifications

CAS Max % 37%, 50%
Packaging Poly Bottle
Quantity 500 mL
UN Number UN1198
Percent Purity 34.5%
Grade Reagent