missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde, 37%, pH 8.6, Reagents

Catalog No. 01336483
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336483 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01336483 Supplier Labchem Inc Supplier No. C72085004F
Only null left

Formaldehyde, 37%, pH 8.6, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 50-00-0 , 50-00-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Formalin, Methanal
IUPAC Name water
SMILES O

Specifications

Boiling Range 96°C
Color Colorless
pH 8.6
Boiling Point 96°C
Packaging Cubitainer
Quantity 4 L
Specific Gravity 1.1
Percent Purity 37%
Grade Reagent
Physical Form Liquid
Chemical Name or Material Formaldehyde, 37%, pH 8.6
Show More Show Less