missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde, 37%, ACS Reagent Grade, Ricca Chemical

Catalog No. rsof0010500
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RSOF0010500 500 mL Natural Poly Bottle
RSOF0010250 250 mL Natural Poly Bottle
RSOF00101A 1 L Natural Poly Bottle
RSOF00104A 4 L Natural Poly Bottle
RSOF001010F 10 L Cubitainer
RSOF001020F 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. RSOF0010500 Supplier Ricca Chemical Company Supplier No. RSOF0010500A
Only null left

Chemical Identifiers

CAS 7732-18-5 , 50-00-0 , 67-56-1
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

CAS Min % 37.0, 47.0, 14.0
CAS Max % 38.0, 49.0, 16.0
Color Clear colorless
Boiling Point Approximately 96°C
Packaging Natural Poly Bottle
Quantity 500 mL
Percent Purity 37% w/w
Grade ACS
Concentration or Composition (by Analyte or Components) 37%
Physical Form Liquid
Chemical Name or Material Formaldehyde, Aqueous (Formalin)
Show More Show Less
ShelfLife 24 months