missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde, 37%, ACS Reagent (Formalin), Reagents

Catalog No. 01337956
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337956 10 L
01337955 1 L
01337954 20 L
01337953 4 L
01337952 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01337956 Supplier Labchem Inc Supplier No. C120850010F

Formaldehyde, 37%, ACS Reagent (Formalin), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 50-00-0 , 67-56-1
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Formalin, Methanal
IUPAC Name water
SMILES O

Specifications

Color Colorless
Boiling Point 96°C
Packaging Cubitainer
Quantity 10 L
Specific Gravity 1.073
Grade ACS
Physical Form Liquid
Chemical Name or Material Formaldehyde, 37%, ACS Reagent (Formalin)