missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde, 26% (w/w), Ricca Chemical

Catalog No. R32110004A
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R32110004A 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R32110004A Supplier Ricca Chemical Company Supplier No. R32110004A
Only null left

Chemical Identifiers

CAS 7732-18-5 , 50-00-0 , 67-56-1
Molecular Formula CH2O
Molecular Weight (g/mol) Mixture
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

CAS Min % 40.27, 58.0
CAS Max % 41.91, 60.0
Color Clear colorless
Boiling Point Approximately 96°C
Packaging Natural Poly Bottle
Quantity 4 L
Percent Purity 26% w/w
Grade Laboratory
Concentration or Composition (by Analyte or Components) 26% (w/w) Aqueous Solution
Physical Form Liquid
Chemical Name or Material Formaldehyde
Show More Show Less
ShelfLife 24 months