missing translation for 'onlineSavingsMsg'
Learn More

Food Blue No. 1 Aluminum Lake, TCI America™

Catalog No. F017925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F017925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F017925G Supplier TCI America Supplier No. F017925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 68921-42-6
Molecular Formula C37H36N2O9S3
Molecular Weight (g/mol) 748.88
MDL Number MFCD00198070
InChI Key CTRXDTYTAAKVSM-UHFFFAOYSA-N
PubChem CID 129894326
IUPAC Name (4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-{ethyl[(3-sulfophenyl)methyl]amino}phenyl)(2-sulfophenyl)methylium
SMILES CCN(CC1=CC=CC(=C1)S(O)(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=CC=C1S(O)(=O)=O

Specifications

Physical Form Powder
Color Blue
Quantity 25 g
Chemical Name or Material Food Blue No. 1 Aluminum Lake