accessibility menu, dialog, popup

UserName

CAS RN 68921-42-6

IUPAC Name: (4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-{ethyl[(3-sulfophenyl)methyl]amino}phenyl)(2-sulfophenyl)methylium
Synonyms: blue 1 lake acid blue 9 aluminum lake
Molecular Weight (g/mol): 748.88
Molecular Formula: C37H36N2O9S3
InChi Key: CTRXDTYTAAKVSM-UHFFFAOYSA-N
SMILES: CCN(CC1=CC=CC(=C1)S(O)(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=CC=C1S(O)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results