missing translation for 'onlineSavingsMsg'
Learn More

Folinic acid calcium salt hydrate, ≥90%, MilliporeSigma™ Supelco™

Folinic acid calcium salt hydrate is a drug, widely used for the treatment of cancer, since it can reduce toxic side effects and helps to improve drug accumulation in tumor tissues.

$227.64 - $1341.78

Specifications

CAS 1492-18-8 (anhydrous)
Physical Form Neat
Linear Formula C20H21 CaN7O7 · xH2 O
Melting Point 240°C to 250°C
Synonym 5-Formyl-5,6,7,8-tetrahydrofolic acid calcium salt; 5-Formyl-5,6,7,8-tetrahydropteroyl-L-glutamic acid calcium salt; 5-HCO-H4 PteGlu; Calcium folinate; Citrovorum factor calcium salt; Leucovorin calcium salt
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111013181
View Documents
Merck Emd Millipore
F7878100MG
100 mg
Each for $227.64
Only null left
 
111013179
View Documents
Merck Emd Millipore
F7878500MG
500 mg
Each for $742.04
Only null left
 
111013180
View Documents
Merck Emd Millipore
F78781G
1 g
Each for $1,341.78
Only null left
 
Description

Description

Featured Industries: Pharmaceutical (small molecule)
Specifications

Specifications

1492-18-8 (anhydrous)
C20H21 CaN7O7 · xH2 O
5-Formyl-5,6,7,8-tetrahydrofolic acid calcium salt; 5-Formyl-5,6,7,8-tetrahydropteroyl-L-glutamic acid calcium salt; 5-HCO-H4 PteGlu; Calcium folinate; Citrovorum factor calcium salt; Leucovorin calcium salt
511.50 (anhydrous basis)
Analytical Standard
Neat
240°C to 250°C
Analytical Standard
≥90%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'